Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(2-thienyl)-1-ethanone, Technical Grade, Thermo Scientific™

Catalog No. CC13005DE
Change view
Click to view available options
Quantity:
250 mg
5 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC13005DE 5 g Amber Glass Bottle
CC13005CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13005DE Supplier Thermo Scientific Chemicals Supplier No. CC13005DE

Chemical Identifiers

CAS 10531-41-6
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
InChI Key UHWNENCHFSDZQP-UHFFFAOYSA-N
Synonym 2-bromo-1-thiophen-2-yl ethanone, 2-2-bromoacetyl thiophene, 2-bromo-1-2-thienyl-1-ethanone, 2-bromoacetyl thiophene, 2-bromo-1-thiophen-2-yl-ethanone, 2-bromo-1-2-thienyl ethanone, 2-bromo-1-thiophen-2-yl ethan-1-one, ethanone, 2-bromo-1-2-thienyl, ethanone, 2-bromo-1-thienyl, 2-bromo-1-2-thienyl ethan-1-one
PubChem CID 2776372
IUPAC Name 2-bromo-1-thiophen-2-ylethanone
SMILES C1=CSC(=C1)C(=O)CBr

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 205.07
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(2-thienyl)-1-ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.