Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1,3-bis(bromomethyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B391325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B391325G 25 g
B39135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B391325G Supplier TCI America Supplier No. B391325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25006-88-6
Molecular Formula C8H7Br3
Molecular Weight (g/mol) 342.86
MDL Number MFCD00229246
InChI Key HSYMWXREIPJGHX-UHFFFAOYSA-N
PubChem CID 621912
IUPAC Name 2-bromo-1,3-bis(bromomethyl)benzene
SMILES BrCC1=CC=CC(CBr)=C1Br

Specifications

Melting Point 106°C
Color White
Quantity 25 g
UN Number 3261
Synonym alpha,alpha′,2-Tribromo-m-xylene
Formula Weight 342.86
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1,3-bis(bromomethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.