Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1,3-thiazol-5-yl]-1-ethanone, ≥90%, Thermo Scientific™

Catalog No. cc19805cb
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19805CB 250 mg
CC19805DA 1 g
CC19805DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC19805CB Supplier Thermo Scientific Chemicals Supplier No. CC19805CB
Only null left
Add to Cart

Chemical Identifiers

CAS 423768-43-8
Molecular Formula C10H8BrN3OS
Molecular Weight (g/mol) 298.158
MDL Number MFCD03407321
InChI Key PQWKERRFBZJRSD-UHFFFAOYSA-N
Synonym 2-bromo-1-4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl thiazol-5-yl ethanone, ethanone,2-bromo-1-4-methyl-2-2-pyrazinyl-5-thiazolyl, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethanone, 5-bromoacetyl-4-methyl-2-pyrazin-2-yl-1,3-thiazole, 2-bromo-1 4-methyl-2-2-pyrazinyl-1,3-thiazol-5-yl-1-ethanone, 2-bromo-1-4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl ethan-1-one
PubChem CID 2776507
IUPAC Name 2-bromo-1-(4-methyl-2-pyrazin-2-yl-1,3-thiazol-5-yl)ethanone
SMILES CC1=C(SC(=N1)C2=NC=CN=C2)C(=O)CBr

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 298.16
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-[4-methyl-2-(2-pyrazinyl)-1, 3-thiazol-5-yl]-1-ethanone
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.