Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(4-phenoxyphenyl)ethanone 95+%, Thermo Scientific™

Catalog No. CC53705CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC53705CB 250 mg
CC53705DA 1 g
CC53705DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC53705CB Supplier Thermo Scientific Chemicals Supplier No. CC53705CB
Add to Cart

Chemical Identifiers

CAS 28179-33-1
Molecular Formula C14H11BrO2
Molecular Weight (g/mol) 291.14
MDL Number MFCD01419855
InChI Key RAXTYMXDSNWNJS-UHFFFAOYSA-N
Synonym 2-bromo-1-4-phenoxyphenyl ethanone, 2-bromo-1-4-phenoxyphenyl ethan-1-one, ethanone,2-bromo-1-4-phenoxyphenyl, 4-phenoxyphenacyl bromide, 2-bromo-4'-phenoxyacetophenone, 4'-phenoxy-2-bromoacetophenone, alpha-bromo-4'-phenoxyacetophenone, ethanone, 2-bromo-1-4-phenoxyphenyl
PubChem CID 14797675
IUPAC Name 2-bromo-1-(4-phenoxyphenyl)ethanone
SMILES BrCC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 291.14
Percent Purity 90%
Chemical Name or Material 2-Bromo-1-(4-phenoxyphenyl)ethanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.