Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-indanol 98.0+%, TCI America™
SDP

Catalog No. B130025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B130025G 25 g
B13005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B130025G Supplier TCI America Supplier No. B130025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5400-80-6
Molecular Formula C9H9BrO
Molecular Weight (g/mol) 213.07
MDL Number MFCD00003798
InChI Key RTESDSDXFLYAKZ-UHFFFAOYNA-N
Synonym 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783
PubChem CID 95444
IUPAC Name 2-bromo-2,3-dihydro-1H-inden-1-ol
SMILES OC1C(Br)CC2=CC=CC=C12

Specifications

Melting Point 131°C
Quantity 25 g
Formula Weight 213.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-1-indanol
EINECSNumber (4)-1771
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.