Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-methylbenzimidazole, 97%

Catalog No. AAH6007503
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6007503 1 g
AAH60075MD 250 mg
AAH6007506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6007503 Supplier Thermo Scientific Chemicals Supplier No. H6007503
Add to Cart

CAS: 49572-60-3 | C8H7BrN2 | 211.06 g/mol

It is a synthetic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthetic building block.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49572-60-3
Molecular Formula C8H7BrN2
Molecular Weight (g/mol) 211.06
MDL Number MFCD00839528
InChI Key PROLKELCEPYEQT-UHFFFAOYSA-N
Synonym 2-bromo-1-methyl-1h-benzo d imidazole, 2-bromo-1-methyl-1h-benzoimidazole, 1h-benzimidazole, 2-bromo-1-methyl, 2-bromo-1-methyl-1h-benzimidazole, 2-bromo-1-methyl-1,3-benzodiazole, 1-methyl-2-bromo-1h-benzimidazole, 2-bromo-1-methyl-1h-1,3-benzodiazole
PubChem CID 797082
SMILES CN1C(Br)=NC2=CC=CC=C12

Specifications

Quantity 1 g
IUPAC Name 2-bromo-1-methyl-1H-1,3-benzodiazole
Formula Weight 211.06
Percent Purity 97%
Chemical Name or Material 2-Bromo-1-methylbenzimidazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.