Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-butene, cis + trans, 98%, stab.

Catalog No. AAA1805814
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1805814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1805814 Supplier Thermo Scientific Chemicals Supplier No. A1805814
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13294-71-8 | C4H7Br | 135.00 g/mol

Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of substituted catechol derivatives and mono-annelated benzenes.

Solubility
Not miscible or difficult to mix in water (664 mg/L 25 EST).

Notes
Store away from oxidizing agents and heat. Store away from electrostatic charges. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13294-71-8
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00000141
InChI Key UILZQFGKPHAAOU-ONEGZZNKSA-N
Synonym 2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene #
PubChem CID 5364387
IUPAC Name (E)-2-bromobut-2-ene
SMILES C\C=C(/C)Br

Specifications

Density 1.328
Boiling Point 82°C to 90°C
Flash Point 1°C (33°F)
Odor Recognizable
Refractive Index 1.461
Quantity 25 g
UN Number UN1993
Beilstein 1733329
Solubility Information Not miscible or difficult to mix in water (664mg/L 25 EST).
Formula Weight 135
Percent Purity 98%
Chemical Name or Material 2-Bromo-2-butene, cis + trans, stabilized
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 236-316-2
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.