Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2'-fluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. B510425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B510425G 25 g
B51045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B510425G Supplier TCI America Supplier No. B510425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 655-15-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.037
MDL Number MFCD00278796
InChI Key QDNWNJSLWKHNTM-UHFFFAOYSA-N
Synonym 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
PubChem CID 2737449
IUPAC Name 2-bromo-1-(2-fluorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)F

Specifications

Color White-Yellow
Boiling Point 63°C
Quantity 25 g
UN Number 3261
Formula Weight 217.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-2′-fluoroacetophenone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.