Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-nitro-1,3-propanediol 98.0+%, TCI America™
SDP

Catalog No. B1247100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1247100G 100 g
B124725G 25 g
B1247500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1247100G Supplier TCI America Supplier No. B1247100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

Melting Point 131°C
Quantity 100 g
UN Number 3241
Formula Weight 199.99
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-2-nitro-1,3-propanediol
EINECSNumber (2)-0325
RTECSNumber TY3385000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.