Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-2-nitropropane-1,3-diol, 98+%

Catalog No. p-7081604
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1163922 100 g
AAA1163936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1163922 Supplier Thermo Scientific Chemicals Supplier No. A1163922
Only null left
Add to Cart

CAS: 52-51-7 | C3H6BrNO4 | 199.988 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Bromo-2-nitropropane-1,3-diol Cosmetics, Preservatives for milk samples, Topical medications and also provide wide spectrum of antibacterial activity.

Solubility
Soluble in water(280g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-51-7
Molecular Formula C3H6BrNO4
Molecular Weight (g/mol) 199.988
MDL Number MFCD00007390
InChI Key LVDKZNITIUWNER-UHFFFAOYSA-N
Synonym bronopol, 2-bromo-2-nitro-1,3-propanediol, bronosol, bronocot, bronidiol, bronopolu, bronotak, 1,3-propanediol, 2-bromo-2-nitro, onyxide 500, lexgard bronopol
PubChem CID 2450
ChEBI CHEBI:31306
IUPAC Name 2-bromo-2-nitropropane-1,3-diol
SMILES C(C(CO)([N+](=O)[O-])Br)O

Specifications

Melting Point ∼122°C to 132°C
pH 5 to 7
Quantity 100 g
UN Number UN3241
Beilstein 1705868
Merck Index 14,1447
Solubility Information Soluble in water(280g/L).
Formula Weight 199.99
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-2-nitropropane-1,3-diol
Hazard Category H302+H312-H315-H318-H335-H500
Hazard Statement GHS H Statement
H301-H331-H318-H228-H312-H315-H335
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Flammable solid.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: 2-BROMO-2-NITROPROPANE-1,3-DIOL
EINECSNumber 200-143-0
RTECSNumber TY3385000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.