Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-methoxynaphthalene 97.0+%, TCI America™
SDP

Catalog No. B34031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34031G 1 g
B34035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34031G Supplier TCI America Supplier No. B34031G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68251-77-4
Molecular Formula C11H9BrO
Molecular Weight (g/mol) 237.10
MDL Number MFCD11870110
InChI Key VFAPZPJQVMNTEX-UHFFFAOYSA-N
PubChem CID 13351910
IUPAC Name 2-bromo-3-methoxynaphthalene
SMILES COC1=C(Br)C=C2C=CC=CC2=C1

Specifications

Melting Point 75°C
Color White-Yellow
Quantity 1 g
Formula Weight 237.10
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-methoxynaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.