Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-methylbutyric Acid 90.0+%, TCI America™
SDP

Catalog No. B0668500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0668500G 500 g
B066825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0668500G Supplier TCI America Supplier No. B0668500G

Chemical Identifiers

CAS 565-74-2
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00004210
InChI Key UEBARDWJXBGYEJ-UHFFFAOYSA-N
Synonym 2-bromo-3-methylbutyric acid, 2-bromoisovaleric acid, butanoic acid, 2-bromo-3-methyl, alpha-bromoisovaleric acid, butyric acid, 2-bromo-3-methyl, 2-bromo-3-methyl-butanoic acid, .alpha.-bromoisovaleric acid, +/--2-bromo-3-methylbutyric acid, bromisovaleriansaure, acmc-209wpb
PubChem CID 11268
IUPAC Name 2-bromo-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)Br

Specifications

Color White-Yellow
Boiling Point 230°C
Quantity 500 g
UN Number 3261
Formula Weight 181.03
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-3-methylbutyric Acid
RTECSNumber ES8055000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.