Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-methylpyridine 97.0+%, TCI America™
SDP

Catalog No. B189425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B189425G 25 g
B18945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B189425G Supplier TCI America Supplier No. B189425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3430-17-9
Molecular Formula C6H6BrN
Molecular Weight (g/mol) 172.03
MDL Number MFCD00239380
InChI Key PZSISEFPCYMBDL-UHFFFAOYSA-N
Synonym 2-bromo-3-picoline, 2-bromo-3-methylpyridin, 2-bromo-3-methyl pyridine, 2-bromo-3-methyl-pyridine, 3-methyl-2-bromopyridine, pyridine, 2-bromo-3-methyl, 2-brom-3-methylpyridin, pubchem9208, 2-bromo-3-mehtylpyridine
PubChem CID 220832
IUPAC Name 2-bromo-3-methylpyridine
SMILES CC1=CC=CN=C1Br

Specifications

Color Yellow
Boiling Point 219°C
Quantity 25 g
Formula Weight 172.03
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.