Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-3-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B332225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B332225G 25 g
B33225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B332225G Supplier TCI America Supplier No. B332225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 573-54-6
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00074899
InChI Key WTDJEGSXLFHZPY-UHFFFAOYSA-N
Synonym 2-bromo-3-nitrobenzoic acid, benzoic acid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoicacid, 2-bromo-3-nitro-benzoic acid, pubchem4714, acmc-1an8y, ksc273m5t, benzoicacid, 2-bromo-3-nitro, 2-bromo-3-nitrobenzoic acid, technical grade
PubChem CID 68452
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])Br)C(=O)O

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 246.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-3-nitrobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.