Learn More
2-Bromo-4,6-dimethylaniline, 98%
Description
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1',1'-dimethylprop-2'-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Bromo-4,6-dimethylaniline can react with 3-chloro-3-methyl-but-1-yne to produce 2-bromo-N-(1′,1′-dimethylprop-2′-ynyl)-4,6-dimethylaniline. This reaction will need reagent Et3N, catalyst CuCl, and the menstruum dioxane.
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 42°C to 46°C |
| Quantity | 25 g |
| UN Number | UN2811 |
| Beilstein | 3046484 |
| Boiling Point | 75°C to 79°C (5 mmHg) |
| Solubility Information | Insoluble in water. |
| Formula Weight | 200.08 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Bromo-4,6-dimethylaniline |
Safety and Handling
| Hazard Category | H302+H312+H332-H315-H319-H373 |
| Hazard Statement | GHS H Statement H331-H373-H302-H312-H315-H319 Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P362-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 246-337-9 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.