Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4,6-dimethylaniline 97.0+%, TCI America™
SDP

Catalog No. B190825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B190825G 25 g
B19085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B190825G Supplier TCI America Supplier No. B190825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 41825-73-4
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00047826
InChI Key YOSJCQJJIHEUKA-UHFFFAOYSA-N
Synonym 2-bromo-4,6-dimethyl-aniline, 6-bromo-2,4-xylidine, benzenamine, 2-bromo-4,6-dimethyl, 2-bromo-4,6-dimethylphenylamine, 4-amino-5-bromo-m-xylene, 2-bromo-4,6-dimethylbenzenamine, 2-bromo-4,6-dimethyl-phenylamine, 2-amino-3,5-dimethyl-1-bromobenzene, pubchem3761
PubChem CID 170579
IUPAC Name 2-bromo-4,6-dimethylaniline
SMILES CC1=CC(C)=C(N)C(Br)=C1

Specifications

Melting Point 45°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 200.08
Percent Purity ≥97.0% (GC,T)
Chemical Name or Material 2-Bromo-4,6-dimethylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.