Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-chloro-1-fluorobenzene 98.0+%, TCI America™
SDP

Catalog No. B295625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B295625G 25 g
B29565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B295625G Supplier TCI America Supplier No. B295625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1996-30-1
Molecular Formula C6H3BrClF
Molecular Weight (g/mol) 209.442
MDL Number MFCD00672934
InChI Key YFFUYGSLQXVHMB-UHFFFAOYSA-N
Synonym 3-bromo-4-fluorochlorobenzene, 3-bromo-1-chloro-4-fluorobenzene, 2-fluoro-5-chlorobromobenzene, 1-bromo-5-chloro-2-fluorobenzene, 1-bromo-3-chloro-6-fluorobenzene, 5-chloro-2-fluorobromobenzene, 2-bromo-4-chloro-1-fluoro-benzene, 2-bromo-4-chlorofluorobenzene, benzene, 2-bromo-4-chloro-1-fluoro, pubchem3210
PubChem CID 2773264
IUPAC Name 2-bromo-4-chloro-1-fluorobenzene
SMILES C1=CC(=C(C=C1Cl)Br)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 25 g
Formula Weight 209.44
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-chloro-1-fluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.