Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-4-chloro-1-iodobenzene, 98%

Click to view available options
Quantity:
1g
5g
25g
31928-44-6
C6H3BrClI
317.348
MFCD00672944
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
2-bromo-4-chloroiodobenzene,benzene, 2-bromo-4-chloro-1-iodo,acmc-209hpo,ksc494q6f,1-iodo-2-bromo-4-chlorobenzene,2-bromo-1-iodo-4-chlorobenzene
13741326
2-bromo-4-chloro-1-iodobenzene
C1=CC(=C(C=C1Cl)Br)I
31928-44-6
C6H3BrClI
317.348
MFCD00672944
CXHXFDQEFKFYQJ-UHFFFAOYSA-N
2-bromo-4-chloroiodobenzene,benzene, 2-bromo-4-chloro-1-iodo,acmc-209hpo,ksc494q6f,1-iodo-2-bromo-4-chlorobenzene,2-bromo-1-iodo-4-chlorobenzene
13741326
2-bromo-4-chloro-1-iodobenzene
C1=CC(=C(C=C1Cl)Br)I
Quantity 25g
Formula Weight 317.36
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloro-1-iodobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning