Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-chloroaniline, 98%, Thermo Scientific™

Catalog No. AAA1412309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1412309 10 g
AAA1412318 50 g
AAA1412330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1412309 Supplier Thermo Scientific Chemicals Supplier No. A1412309

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-38-1
Molecular Formula C6H5BrClN
Molecular Weight (g/mol) 206.467
MDL Number MFCD00041313
InChI Key SYTBIFURTZACKR-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-4-chloro, 1-amino-2-bromo-4-chlorobenzene, 2-bromo-4-chloro-phenylamine, 2-bromo-4-chloro aniline, 2-bromo-4-chlorophenylamine, pubchem3576, acmc-1bkdm, 2-bromo-4-chloro-aniline, 4-chloro-2-bromoaniline, 2-bromo-4-chloroaniline
PubChem CID 70110
IUPAC Name 2-bromo-4-chloroaniline
SMILES C1=CC(=C(C=C1Cl)Br)N

Specifications

Melting Point 65°C to 67°C
Quantity 10 g
Formula Weight 206.48
Percent Purity 98%
Chemical Name or Material 2-Bromo-4-chloroaniline
missing translation for 'healthHazard1' Danger
missing translation for 'healthHazard2' GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.