Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone 98.0+%, TCI America™
SDP

Catalog No. B52131G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B52131G 1 g
B52135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B52131G Supplier TCI America Supplier No. B52131G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 537050-14-9
Molecular Formula C9H5BrF4O
Molecular Weight (g/mol) 285.036
MDL Number MFCD03094509
InChI Key BUNOSKWLVXNWEH-UHFFFAOYSA-N
Synonym 4-fluoro-3-trifluoromethyl phenacyl bromide, 2-bromo-1-4-fluoro-3-trifluoromethyl phenyl ethanone, 4-fluoro-3-trifluoromethyl phenacylbromid, 4-fluoro-3-trifluoromethyl phenacyl bromid, 2-bromo-1-4-fluoro-3-trifluoromethyl phenyl ethan-1-one, 2-bromo-4'-fluoro-3'-trifluoromethyl acetophenone, 4-fluoro-3-trifluoromethyl phenacylbromide, 2-bromo-4'-fluoro-3'-trifluoromethylacetophenone, acmc-209l9v
PubChem CID 2778787
IUPAC Name 2-bromo-1-[4-fluoro-3-(trifluoromethyl)phenyl]ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)C(F)(F)F)F

Specifications

Melting Point 44°C
Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 285.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-fluoro-3′-(trifluoromethyl)acetophenone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.