Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluoroanisole 98.0+%, TCI America™
SDP

Catalog No. B183025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B183025G 25 g
B18305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B183025G Supplier TCI America Supplier No. B183025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-08-4
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00012014
InChI Key JIQXVIJARQLCOY-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoroanisole, benzene, 2-bromo-4-fluoro-1-methoxy, 2-bromo-4-fluoro anisole, 1-bromo-3-fluoro-6-methoxybenzene, 2-bromo-4-fluoro-methoxybenzene, 2-bromo-4-fluoro-1-methoxy-benzene, 1-bromo-5-fluoro-2-methoxybenzene, attercop-chm at106665, pubchem1972, 2-bromo-4-fluoroanisol
PubChem CID 136292
IUPAC Name 2-bromo-4-fluoro-1-methoxybenzene
SMILES COC1=CC=C(F)C=C1Br

Specifications

Color Yellow
Boiling Point 79°C
Quantity 25 g
Formula Weight 205.03
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-4-fluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.