Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B288725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B288725G 25 g
B28875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B288725G Supplier TCI America Supplier No. B288725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1006-41-3
Molecular Formula C7H4BrFO2
Molecular Weight (g/mol) 219.009
MDL Number MFCD00055370
InChI Key RRKPMLZRLKTDQV-UHFFFAOYSA-N
Synonym 2-bromo-4-fluorobenzoicacid, 4-fluoro-2-bromobenzoic acid, 2-bromo-4-fluoro-benzoic acid, benzoic acid, 2-bromo-4-fluoro, rarechem al bo 1996, buttpark 18\02-85, 2-bromo-4-fluorbenzoic acid, pubchem1314, 2-bromo-4-fluorobenzoic, 2-bromofluorobenzoic acid
PubChem CID 1268169
IUPAC Name 2-bromo-4-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)Br)C(=O)O

Specifications

Melting Point 174°C
Color White
Quantity 25 g
Formula Weight 219.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluorobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.