Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorobenzonitrile 98.0+%, TCI America™
SDP

Catalog No. B274425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B274425G 25 g
B27445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B274425G Supplier TCI America Supplier No. B274425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 36282-26-5
Molecular Formula C7H3BrFN
Molecular Weight (g/mol) 200.01
MDL Number MFCD00672924
InChI Key MNNDREXLRLDWEY-UHFFFAOYSA-N
Synonym 4-fluoro-2-bromobenzonitrile, 2-bromo-4-fluoro-benzonitrile, 2-bromo-4-fluorobenzenecarbonitrile, 2-cyano-5-fluorobromobenzene, benzonitrile, 2-bromo-4-fluoro, pubchem3217, acmc-1ajip, ksc490q6h, 2-bromo-4-fluoro benzonitrile, 2-bromo-4-fluorobenzonitrile
PubChem CID 118939
IUPAC Name 2-bromo-4-fluorobenzonitrile
SMILES FC1=CC=C(C#N)C(Br)=C1

Specifications

Melting Point 77°C
Quantity 25 g
UN Number 3439
Formula Weight 200.01
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluorobenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.