Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-fluorophenol 98.0+%, TCI America™
SDP

Catalog No. B155525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B155525G 25 g
B15555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B155525G Supplier TCI America Supplier No. B155525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 496-69-5
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 190.999
MDL Number MFCD00010614
InChI Key MEYRABVEYCFHHB-UHFFFAOYSA-N
Synonym 2-bromo-4-fluoro-phenol, phenol, 2-bromo-4-fluoro, 4-fluoro-2-bromo phenol, pubchem1485, 2-bromo-4-flourophenol, 2-bromo4-fluoro-phenol, acmc-1artu, 2-bromo-4-fluoro phenol, ksc490k9b, akos akm01586
PubChem CID 605472
IUPAC Name 2-bromo-4-fluorophenol
SMILES C1=CC(=C(C=C1F)Br)O

Specifications

Melting Point 43°C
Boiling Point 89°C
Quantity 25 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-fluorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.