Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-methyl-5-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. B506225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B506225G 25 g
B50625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B506225G Supplier TCI America Supplier No. B506225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23056-47-5
Molecular Formula C6H5BrN2O2
Molecular Weight (g/mol) 217.022
MDL Number MFCD03095066
InChI Key OSYUAHGEDKTDOX-UHFFFAOYSA-N
Synonym 2-Bromo-5-nitro-4-picoline
PubChem CID 345873
IUPAC Name 2-bromo-4-methyl-5-nitropyridine
SMILES CC1=CC(=NC=C1[N+](=O)[O-])Br

Specifications

Melting Point 65°C
Color Yellow
Quantity 25 g
Formula Weight 217.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4-methyl-5-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.