Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4'-nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. N0473100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0473100G 100 g
N04735G 5 g
N047325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0473100G Supplier TCI America Supplier No. N0473100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 99-81-0
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.044
MDL Number MFCD00007356
InChI Key MBUPVGIGAMCMBT-UHFFFAOYSA-N
Synonym 2-bromo-4'-nitroacetophenone, 2-bromo-1-4-nitrophenyl ethanone, 4-nitrophenacyl bromide, p-nitrophenacyl bromide, 2-bromo-1-4-nitrophenyl ethan-1-one, alpha-bromo-p-nitroacetophenone, ethanone, 2-bromo-1-4-nitrophenyl, 4'-nitrophenacyl bromide, alpha-bromo-4-nitroacetophenone, 2-bromo-4-nitroacetophenone
PubChem CID 66840
IUPAC Name 2-bromo-1-(4-nitrophenyl)ethanone
SMILES C1=CC(=CC=C1C(=O)CBr)[N+](=O)[O-]

Specifications

Melting Point 98°C
Quantity 100 g
UN Number 1759
Formula Weight 244.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-4′-nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.