Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-4-pyridinemethanol, 96%, Thermo Scientific™

Catalog No. AAL1985603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1985603 1 g
AAL1985606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1985603 Supplier Thermo Scientific Chemicals Supplier No. L1985603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118289-16-0
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.02
MDL Number MFCD04039313
InChI Key IQNUGAFIKDRYRP-UHFFFAOYSA-N
Synonym 2-bromopyridine-4-methanol, 2-bromopyridin-4-yl methanol, 2-bromo-4-hydroxymethylpyridine, 2-bromo-4-hydroxymethyl pyridine, 2-bromo-4-pyridine methanol, 4-pyridinemethanol, 2-bromo, 2-bromo-4-pyridinemethanol, 2-bromo-4-pyridyl methan-1-ol, pubchem7708, 2-bromopyridin-4-methanol
PubChem CID 14512957
IUPAC Name (2-bromopyridin-4-yl)methanol
SMILES OCC1=CC(Br)=NC=C1

Specifications

Melting Point 59°C to 63°C
Quantity 1 g
Beilstein 4244047
Formula Weight 188.02
Percent Purity 96%
Chemical Name or Material 2-Bromo-4-pyridinemethanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.