Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-chlorobenzaldehyde 97.0+%, TCI America™
SDP

Catalog No. B295525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B295525G 25 g
B29555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B295525G Supplier TCI America Supplier No. B295525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 174265-12-4
Molecular Formula C7H4BrClO
Molecular Weight (g/mol) 219.462
InChI Key IIISHLMCTDMUHH-UHFFFAOYSA-N
Synonym 5-chloro-2-bromobenzaldehyde, benzaldehyde, 2-bromo-5-chloro, pubchem19849, acmc-1bwmm, ksc174i5n, 2-bromo-5-chlorobenzaldehycle, iiishlmctdmuhh-uhfffaoysa
PubChem CID 15391279
IUPAC Name 2-bromo-5-chlorobenzaldehyde
SMILES C1=CC(=C(C=C1Cl)C=O)Br

Specifications

Melting Point 66°C
Color White-Yellow
Quantity 25 g
Formula Weight 219.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-chlorobenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.