Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-chlorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. B336625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B336625G 25 g
B33665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B336625G Supplier TCI America Supplier No. B336625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 344-65-0
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00010308
InChI Key OSTIALFVJOFNPP-UHFFFAOYSA-N
Synonym 2-bromo-5-chlorobenzotrifluoride, 1-bromo-4-chloro-2-trifluoromethyl benzene, benzene, 1-bromo-4-chloro-2-trifluoromethyl, 2-bromo-5-chorobenzotrifluoride, 2-bromo-5-chloro-alpha,alpha,alpha-trifluorotoluene, 1-bromo-4-chloro-2-trifluoromethyl-benzene, 4-chloro-2-trifluoromethyl bromobenzene, pubchem2713, bromo-5-chlorobenzotrifluoride
PubChem CID 136172
IUPAC Name 1-bromo-4-chloro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)C(F)(F)F)Br

Specifications

Melting Point 19°C
Color Yellow
Boiling Point 198°C
Quantity 25 g
Formula Weight 259.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-chlorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.