Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-cyanopyridine 97.0+%, TCI America™
SDP

Catalog No. B40391G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40391G 1 g
B40395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40391G Supplier TCI America Supplier No. B40391G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 139585-70-9
Molecular Formula C6H3BrN2
Molecular Weight (g/mol) 183.008
MDL Number MFCD00234141
InChI Key XHYGUDGTUJPSNX-UHFFFAOYSA-N
Synonym 2-bromo-5-cyanopyridine, 6-bromonicotinonitrile, 2-bromo-5-pyridinecarbonitrile, 5-cyano-2-bromopyridine, 6-bromo-nicotinonitrile, 6-bromo-3-pyridinecarbonitrile, 3-pyridinecarbonitrile, 6-bromo, pubchem5963, 2-bromo-5-cyano-pyridine, acmc-209cj9
PubChem CID 5005718
IUPAC Name 6-bromopyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)Br

Specifications

Melting Point 145°C
Quantity 1 g
UN Number 3439
Formula Weight 183.01
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-cyanopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.