Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals

Catalog No. AAH6159603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6159603 1 g
AAH6159606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6159603 Supplier Thermo Scientific Chemicals Supplier No. H6159603

2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-5-fluorobenzyl alcohol is used as intermediates in pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-66-5
Molecular Formula C7H6BrFO
Molecular Weight (g/mol) 205.03
MDL Number MFCD00142876
InChI Key HXGZPMHPSBJMKB-UHFFFAOYSA-N
Synonym 2-bromo-5-fluorobenzyl alcohol, 2-bromo-5-fluorophenyl methanol, 2-bromo-5-fluorobenzylalcohol, benzenemethanol, 2-bromo-5-fluoro, 2-bromo-5-fluorophenyl methan-1-ol, buttpark 87\01-69, 2-bromo-5-fluorbenzyl alcohol, pubchem1963, acmc-1cs63, bromo-5-fluorobenzyl alcohol
PubChem CID 2773349
IUPAC Name (2-bromo-5-fluorophenyl)methanol
SMILES OCC1=C(Br)C=CC(F)=C1

Specifications

Melting Point 91°C to 94°C
Color Yellow
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 205.02
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-fluorobenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.