Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-hydroxypyrazine 95.0+%, TCI America™
SDP

Catalog No. B38811G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38811G 1 g
B3881200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38811G Supplier TCI America Supplier No. B38811G

Chemical Identifiers

CAS 374063-92-0
Molecular Formula C4H3BrN2O
Molecular Weight (g/mol) 174.99
MDL Number MFCD06245330
InChI Key ITTXBHQAWOFJAI-UHFFFAOYSA-N
Synonym 5-bromopyrazin-2-ol, 2-bromo-5-hydroxypyrazine, 5-bromo-2-pyrazinol, 5-bromo-pyrazin-2-ol, 2 1h-pyrazinone, 5-bromo, 5-bromo-2-hydroxypyrazine, 2-bromo-5-hydroxy-pyrazine, pubchem7870, acmc-20a0of, 2-hydroxy-5-bromopyrazine
PubChem CID 2771618
IUPAC Name 5-bromo-1,2-dihydropyrazin-2-one
SMILES BrC1=CNC(=O)C=N1

Specifications

Melting Point 115°C
Quantity 1 g
Formula Weight 174.99
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-hydroxypyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.