Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-iodopyridine 97.0+%, TCI America™
SDP

Catalog No. B376525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B376525G 25 g
B37655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B376525G Supplier TCI America Supplier No. B376525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 73290-22-9
Molecular Formula C5H3BrIN
Molecular Weight (g/mol) 283.89
MDL Number MFCD03095201
InChI Key LLKRSJVPTKFSLS-UHFFFAOYSA-N
Synonym 2-bromo-5-iodo-pyridine, pyridine, 2-bromo-5-iodo, 5-iodo-2-bromopyridine, pubchem5969, 5-iodo-2-bromo-pyridine, acmc-1bb8d, ksc493q0p, 2-bromo-5-iodopyridine
PubChem CID 4738271
IUPAC Name 2-bromo-5-iodopyridine
SMILES BrC1=CC=C(I)C=N1

Specifications

Melting Point 123°C
Color White-Yellow
Quantity 25 g
Formula Weight 283.89
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.