Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-iodotoluene 98.0+%, TCI America™
SDP

Catalog No. B309925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B309925G 25 g
B30995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B309925G Supplier TCI America Supplier No. B309925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 202865-85-8
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.933
MDL Number MFCD00070749
InChI Key QSQOGKONVJDRNH-UHFFFAOYSA-N
PubChem CID 2735574
IUPAC Name 1-bromo-4-iodo-2-methylbenzene
SMILES CC1=C(C=CC(=C1)I)Br

Specifications

Color Yellow
Quantity 25 g
Synonym 2-bromo-5-iodotoluene, benzene,1-bromo-4-iodo-2-methyl, benzene, 1-bromo-4-iodo-2-methyl, pubchem3791, 2-bromo-5-iodo toluene, 2-bromo-5-iodo-toluene, 2-bromo-5-iodotoluene;, acmc-1cq3e, ksc543o8j, 1-bromo-2-methyl-4-iodobenzene
Formula Weight 296.93
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-iodotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.