Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-methylaniline 97.0+%, TCI America™
SDP

Catalog No. B450725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B450725G 25 g
B45075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B450725G Supplier TCI America Supplier No. B450725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53078-85-6
Molecular Formula C7H8BrN
Molecular Weight (g/mol) 186.052
MDL Number MFCD01806278
InChI Key QTAQWOXSUFGGKH-UHFFFAOYSA-N
Synonym 2-bromo-5-methylbenzenamine, 2-bromo-5-methylphenylamine, 2-bromo-5-methyl-phenylamine, benzenamine, 2-bromo-5-methyl, 3-amino-4-bromotoluene, 2-bromo-5-methyl-aniline, 6-bromo-m-toluidine, 2-bromo-5-methyl aniline, pubchem4337, 2-bromo-5-methylbenz
PubChem CID 6422065
IUPAC Name 2-bromo-5-methylaniline
SMILES CC1=CC(=C(C=C1)Br)N

Specifications

Melting Point 33°C
Color White-Yellow
Boiling Point 130°C
Quantity 25 g
Formula Weight 186.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-5-methylaniline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.