Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-n-octylthiophene 97.0+%, TCI America™
SDP

Catalog No. B37385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37385G 5 g
B373825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37385G Supplier TCI America Supplier No. B37385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 172514-63-5
Molecular Formula C12H19BrS
Molecular Weight (g/mol) 275.25
MDL Number MFCD20265580
InChI Key USLMRHXOAXBGCS-UHFFFAOYSA-N
PubChem CID 15440923
IUPAC Name 2-bromo-5-octylthiophene
SMILES CCCCCCCCC1=CC=C(Br)S1

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 112°C
Quantity 5 g
Formula Weight 275.25
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Bromo-5-n-octylthiophene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.