Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitroimidazole, 98%

Catalog No. AAH6115514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6115514 25 g
AAH6115503 1 g
AAH6115506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6115514 Supplier Thermo Scientific Chemicals Supplier No. H6115514

CAS: 65902-59-2 | C3H2BrN3O2 | 191.972 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65902-59-2
Molecular Formula C3H2BrN3O2
Molecular Weight (g/mol) 191.972
MDL Number MFCD09038915
InChI Key UWRJWMLKEHRGOH-UHFFFAOYSA-N
Synonym 2-bromo-4-nitroimidazole, 2-bromo-4-nitro-1h-imidazole, 2-bromo-4-nitro-3h-imidazole, 2-bromo-5-nitroimidazole, 1h-imidazole, 2-bromo-4-nitro, nitrobromoimidazole, pubchem20161, acmc-20aiz1, acmc-1b2vr
PubChem CID 47754
IUPAC Name 2-bromo-5-nitro-1H-imidazole
SMILES C1=C(NC(=N1)Br)[N+](=O)[O-]

Specifications

Quantity 25 g
Formula Weight 191.97
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Bromo-5-nitroimidazole
missing translation for 'healthHazard1' H315-H319-H335
missing translation for 'healthHazard2' GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
missing translation for 'healthHazard3' P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.