Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitrotoluene 98.0+%, TCI America™
SDP

Catalog No. B185325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B185325G 25 g
B18535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B185325G Supplier TCI America Supplier No. B185325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7149-70-4
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007281
InChI Key HIMGPQVBNICCGL-UHFFFAOYSA-N
Synonym 2-bromo-5-nitrotoluene, 4-bromo-3-methylnitrobenzene, benzene, 1-bromo-2-methyl-4-nitro, bromo 5--2-nitrotoluene, 2bromo-5-nitrotoluene, pubchem12553, acmc-209oiv, toluene, 2-bromo-5-nitro, ksc498a8d, methyl 2-bromo-5-nitrobenzene
PubChem CID 251672
IUPAC Name 1-bromo-2-methyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1)[N+](=O)[O-])Br

Specifications

Melting Point 78°C
Boiling Point 143°C
Quantity 25 g
Formula Weight 216.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-5-nitrotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.