Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. B45121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45121G 1 g
B45125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B45121G Supplier TCI America Supplier No. B45121G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49669-14-9
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD12406888
InChI Key ZRXWMMNBTGBCIL-UHFFFAOYSA-N
Synonym 2-bromo-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl-pyridine, pyridine, 2-bromo-6-2-methyl-1,3-dioxolan-2-yl, pubchem16082, ksc496i5p, 2-bromo-6-2-methyl-1,3-dioxolane-2-yl pyridine, 2-bromanyl-6-2-methyl-1,3-dioxolan-2-yl pyridine, 2-bromo-6-2-methyl-1,3 dioxolan-2-yl-pyridine, 6-bromo-2-2'-methyl-1',3'-dioxolan-2'-yl-pyridine
PubChem CID 12164282
IUPAC Name 2-bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
SMILES CC1(OCCO1)C2=NC(=CC=C2)Br

Specifications

Melting Point 45°C
Color Yellow
Quantity 1 g
Formula Weight 244.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.