Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-chloro-4-fluoroaniline 97.0+%, TCI America™
SDP

Catalog No. B368125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B368125G 25 g
B36815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B368125G Supplier TCI America Supplier No. B368125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 201849-14-1
Molecular Formula C6H4BrClFN
Molecular Weight (g/mol) 224.46
MDL Number MFCD00051707
InChI Key LIBGMUMMWYKJSC-UHFFFAOYSA-N
Synonym benzenamine, 2-bromo-6-chloro-4-fluoro, pubchem2922, acmc-209f6h, attercop-chm at130703, 2-bromo-4-fluoro-6-chloroaniline, 2-bromo-6-chloro-4-fluoro-aniline, 2-bromo-6-chloro-4-fluorophenylamine, 6-bromo-2-chloro-4-fluorophenylamine, 2-bromo-6-chloro-4-fluoro-phenylamine, 2-bromo-6-chloro-4-fluoroaniline
PubChem CID 614401
IUPAC Name 2-bromo-6-chloro-4-fluoroaniline
SMILES NC1=C(Cl)C=C(F)C=C1Br

Specifications

Melting Point 57°C
Color Red-White
Quantity 25 g
Formula Weight 224.46
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-chloro-4-fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.