Learn More
Medchemexpress LLC 2-bromo-6-nitrophenol | 13073-25-1 | MFCD02683216 | 98.0% | 218.01 g/mol | C6H4BrNO3 | 100 G

Supplier: Medchemexpress LLC HYW040886100G
2-bromo-6-nitrophenol is an organic intermediate used in chemical synthesis and laboratory research. It is a brominated nitrophenol supplied as a solid for handling and further transformation.
- Cas number: 13073-25-1.
- Molecular formula: C6H4BrNO3.
- Molecular weight: 218.01 g/mol.
- Purity: 98.0%.
- Physical form: solid, light brown to yellow.
- Storage recommendations: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.