Learn More
Medchemexpress LLC 2-bromo-6-nitrophenol | 13073-25-1 | MFCD02683216 | 98.0% | 218.01 g/mol | C6H4BrNO3 | 25 G

Supplier: Medchemexpress LLC 25GHYW040886
2-bromo-6-nitrophenol is a brominated nitrophenol used as a synthetic intermediate in organic research. It is supplied for research use only and is commonly employed in multi-step syntheses and derivatization reactions.
- Cas number 13073-25-1.
- Molecular formula C6H4BrNO3.
- Molecular weight 218.01 g/mol.
- Purity 98.0%.
- Available pack sizes include 1 g, 5 g, 10 g, 25 g, 50 g, and 100 g.
- Intended for research use only; not for human or veterinary use.
- Commonly used as an intermediate in organic synthesis.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.