Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. B220325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B220325G 25 g
B22035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B220325G Supplier TCI America Supplier No. B220325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13073-25-1
Molecular Formula C6H4BrNO3
Molecular Weight (g/mol) 218.01
MDL Number MFCD02683216
InChI Key VEJSIOPQKQXJAT-UHFFFAOYSA-N
Synonym 2-bromo-6-nitrophenol, phenol, 2-bromo-6-nitro, 2-bromo-6-nitro-phenol, 6-bromo-2-nitrophenol, 6-nitro-2-bromophenol, 3-bromo-2-hydroxynitrobenzene, pubchem3114, 2-brom-6-nitrophenol, 2-nitro-6-bromophenol, acmc-209bjb
PubChem CID 13545453
IUPAC Name 2-bromo-6-nitrophenol
SMILES OC1=C(Br)C=CC=C1[N+]([O-])=O

Specifications

Melting Point 68°C
Quantity 25 g
Formula Weight 218.01
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-6-nitrophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.