Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-phenylpyridine 98.0+%, TCI America™
SDP

Catalog No. B51501G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51501G 1 g
B5150200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51501G Supplier TCI America Supplier No. B51501G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 39774-26-0
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.096
MDL Number MFCD00234691
InChI Key XIYPPJVLAAXYAB-UHFFFAOYSA-N
PubChem CID 568679
IUPAC Name 2-bromo-6-phenylpyridine
SMILES C1=CC=C(C=C1)C2=NC(=CC=C2)Br

Specifications

Melting Point 52°C
Color Yellow
Quantity 1 g
Formula Weight 234.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Bromo-6-phenylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.