Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-7-tert-butylpyrene 98.0+%, TCI America™
SDP

Catalog No. B39571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39571G 1 g
B39575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39571G Supplier TCI America Supplier No. B39571G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 78751-80-1
Molecular Formula C20H17Br
Molecular Weight (g/mol) 337.26
InChI Key RYLYZVOQPGVCQT-UHFFFAOYSA-N
PubChem CID 70702341
IUPAC Name 2-bromo-7-tert-butylpyrene
SMILES CC(C)(C)C1=CC2=C3C(=C1)C=CC4=CC(=CC(=C43)C=C2)Br

Specifications

Melting Point 152°C
Color White
Quantity 1 g
Formula Weight 337.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromo-7-tert-butylpyrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.