Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-m-xylene, 98+%

Catalog No. AAA1313022
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
500 g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1313022 100 g
AAA1313009 10 g
AAA1313014 25 g
AAA1313030 250 g
AAA1313036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. AAA1313022 Supplier Thermo Scientific Chemicals Supplier No. A1313022
Add to Cart

CAS: 576-22-7 | C8H9Br | 185.064 g/mol

2-Bromo-m-xylene has been used to prepare 2,2,4,6,6-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromo-m-xylene has been used to prepare 2,2′,4,6,6′-pentamethylbiphenyl. It is used as an intermediate in organic synthesis.

Solubility
Soluble in aqueous solution. Very soluble in ethanol, soluble in acetone and benzene.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents. Should be stored in a tightly closed container. Store in a cool, dry, well-ventilated area.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-22-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000075
InChI Key MYMYVYZLMUEVED-UHFFFAOYSA-N
Synonym 2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206
PubChem CID 68471
IUPAC Name 2-bromo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Br

Specifications

Melting Point -10°C
Density 1.384
Quantity 100 g
Refractive Index 1.555
Beilstein 1929780
Boiling Point 205°C to 206°C
Solubility Information Soluble in aqueous solution. Very soluble in ethanol,soluble in acetone and benzene.
Formula Weight 185.07
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-m-xylene
Hazard Category H227-H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P374-P501c
EINECSNumber 209-397-7
RTECSNumber CY9020000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.