Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-m-xylene 98.0+%, TCI America™
SDP

Catalog No. B0669250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0669250G 250 g
B066925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0669250G Supplier TCI America Supplier No. B0669250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 576-22-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000075
InChI Key MYMYVYZLMUEVED-UHFFFAOYSA-N
Synonym 2,6-dimethylbromobenzene, 2-bromo-m-xylene, 1-bromo-2,6-dimethylbenzene, benzene, 2-bromo-1,3-dimethyl, 2,6-dimethyl bromobenzene, m-xylene, 2-bromo, 2,6-dimethylphenyl bromide, 2-bromo-meta-xylene, 2-bromo-1,3-dimethyl-benzene, pubchem3206
PubChem CID 68471
IUPAC Name 2-bromo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Br

Specifications

Melting Point -10°C
Quantity 250 g
Physical Form Clear Liquid at 20°C
Boiling Point 206°C
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2-Bromo-m-xylene
RTECSNumber CY9020000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.