Learn More
Sigma Organic Chemistry 2-Bromo-N- 2 4-dichloro 1G
Supplier: Sigma Organic Chemistry OTV0001781G

2-Bromo-N- 2 4-dichloro , 1G
About this item:
MDL #: MFCD03014411
Chemical Formula: C10H10BrCl2NO
Molecular Weight: 311
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.