Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoadamantane 97.0+%, TCI America™
SDP

Catalog No. B038025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B038025G 25 g
B03805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B038025G Supplier TCI America Supplier No. B038025G

Chemical Identifiers

CAS 7314-85-4
Molecular Formula C10H15Br
Molecular Weight (g/mol) 215.134
MDL Number MFCD00074745
InChI Key RCXJARRRXOPXBC-UHFFFAOYSA-N
Synonym 2-adamantyl bromide, adamantane, 2-bromo, tricyclo 3.3.1.13,7 decane, 2-bromo, tricyclo 3.3.1.1 3,7 decane, 2-bromo, 1r,3r,5r,7r-2-bromoadamantane, 2-bromo-adamantane, 2-bromanyladamantane, pubchem8752, 2alpha-bromoadamantane, 2-bromotricyclo 3.3.1.13.7 decane
PubChem CID 522482
IUPAC Name 2-bromoadamantane
SMILES C1C2CC3CC1CC(C2)C3Br

Specifications

Melting Point 140°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Bromoadamantane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.