Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzaldehyde, 97%

Catalog No. AC106650250
Change view
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106650250 25 mL Glass bottle
AC106650100 10 mL Glass bottle
AC106651000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC106650250 Supplier Thermo Scientific Chemicals Supplier No. 106650250
Add to Cart

CAS: 6630-33-7 | C7H5BrO | 185.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6630-33-7
Molecular Formula C7H5BrO
Molecular Weight (g/mol) 185.02
MDL Number MFCD00003300
InChI Key NDOPHXWIAZIXPR-UHFFFAOYSA-N
Synonym o-bromobenzaldehyde, benzaldehyde, 2-bromo, benzaldehyde, o-bromo, 2-bromo benzaldehyde, bromobenzaldehyde, bromo benzaldehyde, o-bromo-benzaldehyde, 2-formylbromobenzene, 2-bromo-benzaldehyde, benzaldehyde, bromo
PubChem CID 81129
IUPAC Name 2-bromobenzaldehyde
SMILES BrC1=CC=CC=C1C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 21.0°C to 22.0°C
Color Yellow
Density 1.5800g/mL
Boiling Point 230.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5950 to 1.5970
Linear Formula BrC6H4CHO
Quantity 25 mL
Beilstein 07,238
Specific Gravity 1.58
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 185.02
Percent Purity 97%
Physical Form Liquid After Melting
Chemical Name or Material 2-Bromobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF SWALLOWED: Call a POISO
EINECSNumber 229-622-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.